Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGACCGCCACAGTGAGCACCCAG[A/G]CTCAGGGCACCTGGGGAGCCAGAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 130410 MIM: 154045 MIM: 606008 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLDND2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CLDND2 - claudin domain containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ETFB - electron transfer flavoprotein beta subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LIM2 - lens intrinsic membrane protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NKG7 - natural killer cell granule protein 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005601.3 | 456 | Silent Mutation | AGC,AGT | S,S 150 | NP_005592.1 | |
XM_005258955.3 | 456 | Silent Mutation | AGC,AGT | S,S 115 | XP_005259012.1 | |
XM_006723228.3 | 456 | Silent Mutation | AGC,AGT | S,S 101 | XP_006723291.1 |