Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTTAGGGACTGGGGAGAGAAGAG[A/G]GGTCTGCCCGTCACAAGCATGCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF671 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF671 - zinc finger protein 671 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321375.1 | 2652 | UTR 3 | NP_001308304.1 | |||
NM_001321376.1 | 2652 | UTR 3 | NP_001308305.1 | |||
NM_024833.2 | 2652 | UTR 3 | NP_079109.2 | |||
XM_017027314.1 | 2652 | UTR 3 | XP_016882803.1 |