Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACCCTGAGCTGTGGCGCCTGCTTA[A/T]TAAGCAGGGTGCCTCCATGCTGGAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602216 | ||||||||||||||||||||
Literature Links: |
CBARP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CBARP - CACN beta subunit associated regulatory protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152769.2 | 2285 | UTR 3 | NP_689982.3 | |||
XM_017026555.1 | 2285 | Intron | XP_016882044.1 | |||
XM_017026556.1 | 2285 | Intron | XP_016882045.1 | |||
XM_017026557.1 | 2285 | Intron | XP_016882046.1 | |||
XM_017026558.1 | 2285 | Intron | XP_016882047.1 | |||
XM_017026559.1 | 2285 | UTR 3 | XP_016882048.1 | |||
XM_017026560.1 | 2285 | Intron | XP_016882049.1 | |||
XM_017026561.1 | 2285 | Intron | XP_016882050.1 | |||
XM_017026562.1 | 2285 | Intron | XP_016882051.1 |
STK11 - serine/threonine kinase 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |