Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGCTGATGTTGATGCTGGTGGCGG[C/G]TGTGACCATGTGGCTCCGACCGCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605490 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CATSPERD PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CATSPERD - cation channel sperm associated auxiliary subunit delta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152784.3 | 379 | Missense Mutation | GCT,GGT | A,G 9 | NP_689997.3 | |
XM_011527885.2 | 379 | Missense Mutation | GCT,GGT | A,G 9 | XP_011526187.1 | |
XM_011527886.2 | 379 | Missense Mutation | GCT,GGT | A,G 9 | XP_011526188.1 | |
XM_011527887.2 | 379 | Missense Mutation | GCT,GGT | A,G 9 | XP_011526189.1 | |
XM_011527888.2 | 379 | Missense Mutation | GCT,GGT | A,G 9 | XP_011526190.1 | |
XM_011527889.2 | 379 | Missense Mutation | GCT,GGT | A,G 9 | XP_011526191.1 | |
XM_011527890.2 | 379 | Missense Mutation | GCT,GGT | A,G 9 | XP_011526192.1 | |
XM_011527891.1 | 379 | Intron | XP_011526193.1 | |||
XM_011527892.1 | 379 | Intron | XP_011526194.1 | |||
XM_017026564.1 | 379 | Missense Mutation | GCT,GGT | A,G 9 | XP_016882053.1 | |
XM_017026565.1 | 379 | UTR 5 | XP_016882054.1 | |||
XM_017026566.1 | 379 | Missense Mutation | GCT,GGT | A,G 9 | XP_016882055.1 |
LONP1 - lon peptidase 1, mitochondrial | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001276479.1 | 379 | Intron | NP_001263408.1 | |||
NM_001276480.1 | 379 | Intron | NP_001263409.1 | |||
NM_004793.3 | 379 | Intron | NP_004784.2 | |||
XM_011528441.2 | 379 | Intron | XP_011526743.1 |