Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGCCACTGCTTCCCTGAAACCAT[C/T]GTTCCCACGTCACCGATGGCCCCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606919 MIM: 602880 MIM: 601430 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CERS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CERS1 - ceramide synthase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001290265.1 | 1749 | UTR 3 | NP_001277194.1 | |||
NM_021267.4 | 1749 | Intron | NP_067090.1 | |||
NM_198207.2 | 1749 | UTR 3 | NP_937850.1 |
GDF1 - growth differentiation factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001492.5 | 1749 | Intron | NP_001483.3 |
UPF1 - UPF1, RNA helicase and ATPase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |