Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAACCGTAGAGACTTGGCAGAATA[A/G]CTCTTTAAACTTACAAATAACTTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603971 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF91 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF91 - zinc finger protein 91 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300951.1 | Intron | NP_001287880.1 | ||||
NM_003430.3 | Intron | NP_003421.2 | ||||
XM_006722880.3 | Intron | XP_006722943.1 | ||||
XM_011528265.2 | Intron | XP_011526567.1 | ||||
XM_011528266.2 | Intron | XP_011526568.1 | ||||
XM_017027235.1 | Intron | XP_016882724.1 | ||||
XM_017027236.1 | Intron | XP_016882725.1 |