Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGTTACTTATCAACAAAATAACT[G/T]TCCATCCGGTGGGGAAGTGCCATCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606016 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KEAP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KEAP1 - kelch like ECH associated protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012289.3 | 2655 | UTR 3 | NP_036421.2 | |||
NM_203500.1 | 2655 | UTR 3 | NP_987096.1 | |||
XM_005260173.1 | 2655 | UTR 3 | XP_005260230.1 | |||
XM_005260174.1 | 2655 | UTR 3 | XP_005260231.1 | |||
XM_011528452.1 | 2655 | UTR 3 | XP_011526754.1 |