Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTTGGCTTTTGTCATTGTTTCCC[A/G]GTTATCTTGATTATACATCATACAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF831 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF831 - zinc finger protein 831 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178457.2 | Intron | NP_848552.1 | ||||
XM_005260272.3 | Intron | XP_005260329.1 | ||||
XM_005260273.3 | Intron | XP_005260330.1 | ||||
XM_006723698.3 | Intron | XP_006723761.1 | ||||
XM_011528534.2 | Intron | XP_011526836.1 | ||||
XM_011528536.2 | Intron | XP_011526838.1 | ||||
XM_011528537.2 | Intron | XP_011526839.1 | ||||
XM_011528538.2 | Intron | XP_011526840.1 | ||||
XM_011528540.2 | Intron | XP_011526842.1 | ||||
XM_017027642.1 | Intron | XP_016883131.1 | ||||
XM_017027643.1 | Intron | XP_016883132.1 | ||||
XM_017027644.1 | Intron | XP_016883133.1 |