Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCACGATTACGCAGACAGGAACA[C/T]TGCCTACTTCTGGAATCGCTTCAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604205 MIM: 603038 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CPNE1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CPNE1 - copine 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPAG4 - sperm associated antigen 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317931.1 | 611 | Missense Mutation | ACT,ATT | T,I 205 | NP_001304860.1 | |
NM_003116.2 | 611 | Missense Mutation | ACT,ATT | T,I 282 | NP_003107.1 | |
XM_005260520.4 | 611 | Missense Mutation | ACT,ATT | T,I 189 | XP_005260577.1 | |
XM_011529009.2 | 611 | Missense Mutation | ACT,ATT | T,I 238 | XP_011527311.1 |