Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGCTGGGCTTGGTGGTGAAGGCGG[C/T]GCACGGGCTCCTCCAGCCCGAAGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608188 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CRLS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CRLS1 - cardiolipin synthase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LRRN4 - leucine rich repeat neuronal 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152611.4 | 1250 | Missense Mutation | ACC,GCC | T,A 672 | NP_689824.2 | |
XM_011529183.2 | 1250 | Missense Mutation | ACC,GCC | T,A 335 | XP_011527485.1 |