Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAAAAACTCCCCTTGACAATAAAC[A/G]TAAGTCCAAGCCACAGAGGGGAGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609018 MIM: 600892 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HLCS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HLCS - holocarboxylase synthetase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000411.6 | 2769 | UTR 3 | NP_000402.3 | |||
NM_001242784.1 | 2769 | UTR 3 | NP_001229713.1 | |||
NM_001242785.1 | 2769 | UTR 3 | NP_001229714.1 | |||
XM_005260953.3 | 2769 | UTR 3 | XP_005261010.1 | |||
XM_005260955.3 | 2769 | UTR 3 | XP_005261012.1 | |||
XM_005260956.3 | 2769 | UTR 3 | XP_005261013.1 | |||
XM_006723994.2 | 2769 | UTR 3 | XP_006724057.1 | |||
XM_006723995.1 | 2769 | UTR 3 | XP_006724058.1 | |||
XM_011529538.1 | 2769 | UTR 3 | XP_011527840.1 | |||
XM_011529539.2 | 2769 | UTR 3 | XP_011527841.1 | |||
XM_011529540.2 | 2769 | Intron | XP_011527842.1 | |||
XM_011529541.2 | 2769 | UTR 3 | XP_011527843.1 | |||
XM_017028330.1 | 2769 | UTR 3 | XP_016883819.1 |
SIM2 - single-minded family bHLH transcription factor 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |