Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCAGTCGCAGGAAGCTCTAACGGC[A/G]AAGGAGCACCCGGTGCGGGTGGGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605298 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DSCR3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DSCR3 - DSCR3 arrestin fold containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006052.1 | Intron | NP_006043.1 | ||||
XM_005260909.4 | Intron | XP_005260966.1 | ||||
XM_005260911.4 | Intron | XP_005260968.1 | ||||
XM_011529423.2 | Intron | XP_011527725.1 |
DSCR9 - Down syndrome critical region 9 (non-protein coding) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |