Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATATTATATATATTTGATTTTCA[A/G]TTGCTTTGGTAAAAAGATAACTATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606481 MIM: 610918 MIM: 602127 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
INPP5J PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
INPP5J - inositol polyphosphate-5-phosphatase J | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002837.2 | Intron | NP_001002837.1 | ||||
NM_001284285.1 | Intron | NP_001271214.1 | ||||
NM_001284286.1 | Intron | NP_001271215.1 | ||||
NM_001284287.1 | Intron | NP_001271216.1 | ||||
NM_001284288.1 | Intron | NP_001271217.1 | ||||
NM_001284289.1 | Intron | NP_001271218.1 | ||||
XM_011530143.1 | Intron | XP_011528445.1 | ||||
XM_017028771.1 | Intron | XP_016884260.1 | ||||
XM_017028772.1 | Intron | XP_016884261.1 | ||||
XM_017028773.1 | Intron | XP_016884262.1 |
SELM - selenoprotein M | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SMTN - smoothelin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |