Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAAGAGAACGAGCGTCCCAGCAAC[G/A]GAGGAGGCCAGGAGAGGTAGGGGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601786 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CSDC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CSDC2 - cold shock domain containing C2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PMM1 - phosphomannomutase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002676.2 | Intron | NP_002667.2 | ||||
XM_005261638.4 | Intron | XP_005261695.1 | ||||
XM_011530229.1 | Intron | XP_011528531.1 | ||||
XM_011530230.2 | Intron | XP_011528532.1 | ||||
XM_011530231.2 | Intron | XP_011528533.1 | ||||
XM_011530232.1 | Intron | XP_011528534.1 |