Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCAGTTCTCCAGCATCCTTTTTT[G/T]CCCTCGCTGTCTGGCCTCCGATATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607295 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MYO18B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MYO18B - myosin XVIIIB | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318245.1 | Intron | NP_001305174.1 | ||||
NM_032608.6 | Intron | NP_115997.5 | ||||
XM_011530458.2 | Intron | XP_011528760.1 | ||||
XM_011530459.2 | Intron | XP_011528761.1 | ||||
XM_011530460.2 | Intron | XP_011528762.1 | ||||
XM_011530461.2 | Intron | XP_011528763.1 | ||||
XM_011530464.2 | Intron | XP_011528766.1 | ||||
XM_011530465.2 | Intron | XP_011528767.1 | ||||
XM_011530466.2 | Intron | XP_011528768.1 | ||||
XM_017029012.1 | Intron | XP_016884501.1 | ||||
XM_017029013.1 | Intron | XP_016884502.1 | ||||
XM_017029014.1 | Intron | XP_016884503.1 | ||||
XM_017029015.1 | Intron | XP_016884504.1 | ||||
XM_017029016.1 | Intron | XP_016884505.1 | ||||
XM_017029017.1 | Intron | XP_016884506.1 |