Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGGGGTGGCGGCCATGTGGAGCA[A/G]ACTGAGGGTGTCCTCCAGCCCCCCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300522 MIM: 314690 | ||||||||||||||||||||
Literature Links: |
IQSEC2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IQSEC2 - IQ motif and Sec7 domain 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001111125.2 | 4255 | Missense Mutation | TCT,TTT | S,F 1353 | NP_001104595.1 | |
NM_001243197.1 | 4255 | Intron | NP_001230126.1 | |||
NM_015075.1 | 4255 | UTR 3 | NP_055890.1 | |||
XM_006724579.2 | 4255 | Missense Mutation | TCT,TTT | S,F 1385 | XP_006724642.1 | |
XM_006724580.3 | 4255 | Missense Mutation | TCT,TTT | S,F 1148 | XP_006724643.1 | |
XM_006724581.3 | 4255 | Intron | XP_006724644.1 | |||
XM_006724582.3 | 4255 | Intron | XP_006724645.1 | |||
XM_006724583.3 | 4255 | Intron | XP_006724646.1 | |||
XM_006724584.2 | 4255 | UTR 3 | XP_006724647.1 | |||
XM_011530773.2 | 4255 | Missense Mutation | TCT,TTT | S,F 1116 | XP_011529075.1 | |
XM_011530774.2 | 4255 | Intron | XP_011529076.1 | |||
XM_011530776.1 | 4255 | Intron | XP_011529078.1 | |||
XM_011530777.1 | 4255 | Intron | XP_011529079.1 | |||
XM_017029359.1 | 4255 | Missense Mutation | TCT,TTT | S,F 1343 | XP_016884848.1 | |
XM_017029360.1 | 4255 | Missense Mutation | TCT,TTT | S,F 1187 | XP_016884849.1 |
KDM5C - lysine demethylase 5C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |