Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATGGTCCCATTCCATAGGCTGTG[A/G]AGCTGAATCGGCTTTCCCTTACCTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600421 | ||||||||||||||||||||
Literature Links: |
GLRA3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GLRA3 - glycine receptor alpha 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042543.2 | 1652 | Missense Mutation | TCC,TTC | S,F 368 | NP_001036008.1 | |
NM_006529.3 | 1652 | Missense Mutation | TCC,TTC | S,F 383 | NP_006520.2 | |
XM_011532267.2 | 1652 | Intron | XP_011530569.1 | |||
XM_011532268.2 | 1652 | Missense Mutation | TCC,TTC | S,F 188 | XP_011530570.1 | |
XM_017008630.1 | 1652 | Missense Mutation | TCC,TTC | S,F 343 | XP_016864119.1 |