Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACTGCCAAGGCGCTCTCCGATGA[C/T]GAGCAGGGTGGAAGCGCCTTCGTAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRIML1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TRIML1 - tripartite motif family like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178556.3 | 439 | Silent Mutation | GAC,GAT | D,D 108 | NP_848651.2 | |
XM_006714207.3 | 439 | Silent Mutation | GAC,GAT | D,D 108 | XP_006714270.1 | |
XM_011531919.1 | 439 | Silent Mutation | GAC,GAT | D,D 11 | XP_011530221.1 |