Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGATTGCTGCAAGGCTGAGGGACG[A/C]GAACGAGGTCAGAGCGCTTCTCTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612839 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TET2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TET2 - tet methylcytosine dioxygenase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127208.2 | 310 | UTR 5 | NP_001120680.1 | |||
NM_017628.4 | 310 | Intron | NP_060098.3 | |||
XM_005263082.2 | 310 | UTR 5 | XP_005263139.1 | |||
XM_006714242.3 | 310 | UTR 5 | XP_006714305.1 | |||
XM_017008319.1 | 310 | UTR 5 | XP_016863808.1 |