Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGAAAAAATGTCTATTGGGCCTAA[C/T]GGCAAAGATGTCATAATTGACTTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C4orf22 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C4orf22 - chromosome 4 open reading frame 22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206997.1 | Intron | NP_001193926.1 | ||||
NM_152770.2 | Intron | NP_689983.2 | ||||
XM_011531816.2 | Intron | XP_011530118.2 | ||||
XM_017007972.1 | Intron | XP_016863461.1 | ||||
XM_017007973.1 | Intron | XP_016863462.1 | ||||
XM_017007974.1 | Intron | XP_016863463.1 | ||||
XM_017007975.1 | Intron | XP_016863464.1 | ||||
XM_017007976.1 | Intron | XP_016863465.1 | ||||
XM_017007977.1 | Intron | XP_016863466.1 | ||||
XM_017007978.1 | Intron | XP_016863467.1 |