Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATTGTAACATTATGCTATTATTAA[C/T]GATGAGATGTTTTGTTTTTGTATTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TTC29 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TTC29 - tetratricopeptide repeat domain 29 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300761.2 | Intron | NP_001287690.1 | ||||
NM_001317806.1 | Intron | NP_001304735.1 | ||||
NM_031956.3 | Intron | NP_114162.2 | ||||
XM_005263270.1 | Intron | XP_005263327.1 | ||||
XM_006714335.1 | Intron | XP_006714398.1 | ||||
XM_006714336.1 | Intron | XP_006714399.1 | ||||
XM_006714339.2 | Intron | XP_006714402.1 | ||||
XM_011532310.2 | Intron | XP_011530612.1 |