Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGATAAACAGAAACCGACAACCT[C/T]GTACATAGACCTATTCAAAATCAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
![]() |
||||||||||||||||||||||||||||||||||||||
Literature Links: |
SEC31A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SEC31A - SEC31 homolog A, COPII coat complex component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077206.3 | Intron | NP_001070674.1 | ||||
NM_001077207.3 | Intron | NP_001070675.1 | ||||
NM_001077208.3 | Intron | NP_001070676.1 | ||||
NM_001191049.2 | Intron | NP_001177978.1 | ||||
NM_001300744.2 | Intron | NP_001287673.1 | ||||
NM_001300745.2 | Intron | NP_001287674.1 | ||||
NM_001318119.1 | Intron | NP_001305048.1 | ||||
NM_001318120.1 | Intron | NP_001305049.1 | ||||
NM_016211.4 | Intron | NP_057295.2 |