Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGATAATTATCTGCTTATTTTAA[C/T]AAACCAGCTCTCTAGTTTTAATCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
FSTL5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FSTL5 - follistatin like 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128427.2 | 4308 | UTR 3 | NP_001121899.1 | |||
NM_001128428.2 | 4308 | UTR 3 | NP_001121900.1 | |||
NM_020116.4 | 4308 | UTR 3 | NP_064501.2 | |||
XM_011532126.1 | 4308 | UTR 3 | XP_011530428.1 |