Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGGTCGGAGGCCCCAAGGTCACC[A/G]GGGCCACTGCCCGTCACAGGTCCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
13 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605851 MIM: 616923 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ICMT PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ICMT - isoprenylcysteine carboxyl methyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012405.3 | 642 | UTR 3 | NP_036537.1 | |||
XM_011541140.2 | 642 | UTR 3 | XP_011539442.1 |
RNF207 - ring finger protein 207 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |