Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACTAATGGTGGTCAAGGTGGCTCA[A/G]GTGCCATTGGTGGTGGTCCACAGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
16 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614269 MIM: 609971 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPR153 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GPR153 - G protein-coupled receptor 153 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207370.2 | 3004 | UTR 3 | NP_997253.2 | |||
XM_011541434.2 | 3004 | UTR 3 | XP_011539736.1 | |||
XM_017001250.1 | 3004 | UTR 3 | XP_016856739.1 | |||
XM_017001251.1 | 3004 | Intron | XP_016856740.1 |
HES3 - hes family bHLH transcription factor 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LINC00337 - long intergenic non-protein coding RNA 337 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |