Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGCGCACTGGCCACAGCACTACCC[C/T]GCCTGTGGGGGCCAGAGACAGTCGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
14 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 114780 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CA6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CA6 - carbonic anhydrase 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001215.3 | 221 | Silent Mutation | CCC,CCT | P,P 40 | NP_001206.2 | |
NM_001270500.1 | 221 | Silent Mutation | CCC,CCT | P,P 40 | NP_001257429.1 | |
NM_001270501.1 | 221 | Intron | NP_001257430.1 | |||
NM_001270502.1 | 221 | Intron | NP_001257431.1 | |||
XM_011542083.2 | 221 | Silent Mutation | CCC,CCT | P,P 44 | XP_011540385.1 | |
XM_011542084.2 | 221 | Silent Mutation | CCC,CCT | P,P 44 | XP_011540386.1 |