Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCGCCATTCCGCCACGGCGCCGA[A/C]AGTCGTCATCAAGCTGCGCGCAGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613450 MIM: 610457 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MZT2B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MZT2B - mitotic spindle organizing protein 2B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005263795.4 | Intron | XP_005263852.1 | ||||
XM_011511886.2 | Intron | XP_011510188.1 | ||||
XM_011511887.2 | Intron | XP_011510189.1 | ||||
XM_011511888.2 | Intron | XP_011510190.1 | ||||
XM_011511889.2 | Intron | XP_011510191.1 | ||||
XM_011511890.2 | Intron | XP_011510192.1 |
SMPD4 - sphingomyelin phosphodiesterase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171083.2 | Intron | NP_001164554.1 | ||||
NM_017751.4 | Intron | NP_060221.2 | ||||
NM_017951.4 | Intron | NP_060421.2 | ||||
XM_011511443.2 | Intron | XP_011509745.1 | ||||
XM_011511444.2 | Intron | XP_011509746.1 | ||||
XM_011511445.2 | Intron | XP_011509747.1 | ||||
XM_017004448.1 | Intron | XP_016859937.1 | ||||
XM_017004449.1 | Intron | XP_016859938.1 |