Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCCTTTAAACTGCAGGACTGCAA[C/T]TGATCTCTGTATGTTTTCTGGTTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608209 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DPP10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DPP10 - dipeptidyl peptidase like 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004360.3 | 20 | Intron | NP_001004360.2 | |||
NM_001178034.1 | 20 | Intron | NP_001171505.1 | |||
NM_001178036.1 | 20 | Intron | NP_001171507.1 | |||
NM_001178037.1 | 20 | Intron | NP_001171508.1 | |||
NM_001321905.1 | 20 | Intron | NP_001308834.1 | |||
NM_001321906.1 | 20 | Intron | NP_001308835.1 | |||
NM_001321907.1 | 20 | UTR 5 | NP_001308836.1 | |||
NM_001321908.1 | 20 | Intron | NP_001308837.1 | |||
NM_001321909.1 | 20 | Intron | NP_001308838.1 | |||
NM_001321910.1 | 20 | UTR 5 | NP_001308839.1 | |||
NM_001321911.1 | 20 | Intron | NP_001308840.1 | |||
NM_001321912.1 | 20 | Intron | NP_001308841.1 | |||
NM_001321913.1 | 20 | UTR 5 | NP_001308842.1 | |||
NM_001321914.1 | 20 | Intron | NP_001308843.1 | |||
NM_020868.4 | 20 | UTR 5 | NP_065919.2 | |||
XM_017004565.1 | 20 | Intron | XP_016860054.1 | |||
XM_017004566.1 | 20 | Intron | XP_016860055.1 |