Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGGGCCCTGCCGGCCCCGCGTGA[G/T]GCCCAGCAAGGCCCGGCTCTGGTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604521 MIM: 609050 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HAAO PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HAAO - 3-hydroxyanthranilate 3,4-dioxygenase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MTA3 - metastasis associated 1 family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282755.1 | 1196 | Intron | NP_001269684.1 | |||
NM_001282756.1 | 1196 | Intron | NP_001269685.1 | |||
NM_020744.3 | 1196 | Intron | NP_065795.1 | |||
XM_005264456.2 | 1196 | Intron | XP_005264513.1 | |||
XM_005264458.2 | 1196 | Intron | XP_005264515.1 | |||
XM_005264459.2 | 1196 | Intron | XP_005264516.1 | |||
XM_017004561.1 | 1196 | Intron | XP_016860050.1 | |||
XM_017004562.1 | 1196 | Intron | XP_016860051.1 |
OXER1 - oxoeicosanoid receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_148962.4 | 1196 | Missense Mutation | ATC,CTC | I,L 372 | NP_683765.1 |