Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACATACAGCTAGTAAGAGGCAGAG[A/G]TGAAGCATTCACAAGTAGTTTGGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615427 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ELMOD3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ELMOD3 - ELMO domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135021.1 | Intron | NP_001128493.1 | ||||
NM_001135022.1 | Intron | NP_001128494.1 | ||||
NM_001135023.1 | Intron | NP_001128495.1 | ||||
NM_032213.4 | Intron | NP_115589.2 | ||||
XM_005264596.3 | Intron | XP_005264653.1 | ||||
XM_005264597.3 | Intron | XP_005264654.1 | ||||
XM_005264598.3 | Intron | XP_005264655.1 | ||||
XM_017005078.1 | Intron | XP_016860567.1 | ||||
XM_017005079.1 | Intron | XP_016860568.1 |