Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGAAATGATACCGAAACGATGCC[A/G]TGATGTGGTTGGCTCTCTAGGCAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 153615 MIM: 615427 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAPG PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CAPG - capping actin protein, gelsolin like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256139.1 | Intron | NP_001243068.1 | ||||
NM_001256140.1 | Intron | NP_001243069.1 | ||||
NM_001320732.1 | Intron | NP_001307661.1 | ||||
NM_001320733.1 | Intron | NP_001307662.1 | ||||
NM_001320734.1 | Intron | NP_001307663.1 | ||||
NM_001747.3 | Intron | NP_001738.2 | ||||
XM_011533122.1 | Intron | XP_011531424.1 | ||||
XM_011533123.1 | Intron | XP_011531425.1 |
ELMOD3 - ELMO domain containing 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |