Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGTATAATGGTTTGCTCCAACAG[C/T]GAGGGGAATAGGAAGGCTCTTTGTA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 602430 | |||||||||||||||||||||||
Literature Links: |
ROBO1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
ROBO1 - roundabout guidance receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145845.1 | 5333 | UTR 3 | NP_001139317.1 | |||
NM_002941.3 | 5333 | UTR 3 | NP_002932.1 | |||
NM_133631.3 | 5333 | UTR 3 | NP_598334.2 | |||
XM_006713277.2 | 5333 | UTR 3 | XP_006713340.1 | |||
XM_011533976.1 | 5333 | UTR 3 | XP_011532278.1 | |||
XM_011533977.2 | 5333 | UTR 3 | XP_011532279.1 | |||
XM_011533978.1 | 5333 | UTR 3 | XP_011532280.1 | |||
XM_011533979.1 | 5333 | UTR 3 | XP_011532281.1 | |||
XM_011533980.1 | 5333 | UTR 3 | XP_011532282.1 | |||
XM_017006982.1 | 5333 | UTR 3 | XP_016862471.1 | |||
XM_017006983.1 | 5333 | UTR 3 | XP_016862472.1 | |||
XM_017006984.1 | 5333 | UTR 3 | XP_016862473.1 | |||
XM_017006985.1 | 5333 | UTR 3 | XP_016862474.1 |