Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAATGTACTAAATGGCTCCCTCGTG[A/C]TCTGAAAATGTTAACACGGTCACAG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 613167 | |||||||||||||||||||||||
Literature Links: |
DCLK3 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
DCLK3 - doublecortin like kinase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033403.1 | 3163 | UTR 3 | NP_208382.1 | |||
XM_011534167.2 | 3163 | UTR 3 | XP_011532469.1 | |||
XM_011534168.2 | 3163 | UTR 3 | XP_011532470.1 | |||
XM_011534169.1 | 3163 | Intron | XP_011532471.1 |