Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCATATGCTAAATAGGATTGTCAG[C/G]AGCTGAAGAGGTCCTGGGGGTGATT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 600163 | |||||||||||||||||||||||
Literature Links: |
SCN5A PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
SCN5A - sodium voltage-gated channel alpha subunit 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000335.4 | 7605 | UTR 3 | NP_000326.2 | |||
NM_001099404.1 | 7605 | UTR 3 | NP_001092874.1 | |||
NM_001099405.1 | 7605 | UTR 3 | NP_001092875.1 | |||
NM_001160160.1 | 7605 | UTR 3 | NP_001153632.1 | |||
NM_001160161.1 | 7605 | UTR 3 | NP_001153633.1 | |||
NM_198056.2 | 7605 | UTR 3 | NP_932173.1 | |||
XM_011533991.2 | 7605 | UTR 3 | XP_011532293.1 | |||
XM_017007017.1 | 7605 | UTR 3 | XP_016862506.1 |