Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAGGAAAATATGATTTTTTGGGGC[A/C]AAACTGATTTATTTGTGCTATAGCT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
ZNF621 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
ZNF621 - zinc finger protein 621 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098414.2 | Intron | NP_001091884.1 | ||||
NM_001287245.1 | Intron | NP_001274174.1 | ||||
NM_198484.4 | Intron | NP_940886.1 | ||||
XM_005265079.3 | Intron | XP_005265136.1 | ||||
XM_005265080.3 | Intron | XP_005265137.1 | ||||
XM_006713114.3 | Intron | XP_006713177.1 | ||||
XM_011533610.2 | Intron | XP_011531912.1 | ||||
XM_017006226.1 | Intron | XP_016861715.1 |