Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCGGGAGGCAGATGGGCTGCACG[C/T]ACTCACTGAAGGTGACAGGTGTTTG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
PRSS43 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
PRSS43 - protease, serine 43 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRSS45 - protease, serine 45 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_199183.2 | Intron | NP_954652.2 |
PRSS46 - protease, serine 46 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |