Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGTCAGCAGGAAAAAGATCACAA[C/G]CCTGCCACGATGGCTGCTGCTCCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605458 | ||||||||||||||||||||
Literature Links: |
ACTR8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ACTR8 - ARP8 actin-related protein 8 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022899.4 | 1388 | Intron | NP_075050.3 | |||
XM_005265587.4 | 1388 | Intron | XP_005265644.1 | |||
XM_011534249.2 | 1388 | Intron | XP_011532551.1 |
IL17RB - interleukin 17 receptor B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018725.3 | 1388 | Missense Mutation | CCC,GCC | P,A 494 | NP_061195.2 | |
XM_005265310.4 | 1388 | Missense Mutation | CCC,GCC | P,A 523 | XP_005265367.1 | |
XM_005265311.4 | 1388 | Missense Mutation | CCC,GCC | P,A 507 | XP_005265368.1 | |
XM_005265312.4 | 1388 | Missense Mutation | CCC,GCC | P,A 478 | XP_005265369.1 | |
XM_011533941.2 | 1388 | UTR 3 | XP_011532243.1 | |||
XM_017006804.1 | 1388 | Missense Mutation | CCC,GCC | P,A 406 | XP_016862293.1 | |
XM_017006805.1 | 1388 | Missense Mutation | CCC,GCC | P,A 390 | XP_016862294.1 | |
XM_017006806.1 | 1388 | Missense Mutation | CCC,GCC | P,A 377 | XP_016862295.1 | |
XM_017006807.1 | 1388 | UTR 3 | XP_016862296.1 |