Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCCAAGTTGATCCCTTACAGCTT[A/C]AAACACCGCCTCAGACACAACCAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614843 | ||||||||||||||||||||
Literature Links: |
ODAM PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ODAM - odontogenic, ameloblast asssociated | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017855.3 | 357 | Missense Mutation | AAA,CAA | K,Q 114 | NP_060325.3 | |
XM_005265698.3 | 357 | Missense Mutation | AAA,CAA | K,Q 131 | XP_005265755.1 | |
XM_006714245.3 | 357 | Missense Mutation | AAA,CAA | K,Q 131 | XP_006714308.1 | |
XM_017008331.1 | 357 | Missense Mutation | AAA,CAA | K,Q 100 | XP_016863820.1 |