Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTATTAGGGTTCTCCATAGAAACC[C/T]AGCGATTAGGAGAGATGATGTATTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614365 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AASDH PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AASDH - aminoadipate-semialdehyde dehydrogenase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286668.1 | 3983 | Intron | NP_001273597.1 | |||
NM_001286669.1 | 3983 | Intron | NP_001273598.1 | |||
NM_001286670.1 | 3983 | Intron | NP_001273599.1 | |||
NM_001286671.1 | 3983 | Intron | NP_001273600.1 | |||
NM_001286672.1 | 3983 | Intron | NP_001273601.1 | |||
NM_001323890.1 | 3983 | Intron | NP_001310819.1 | |||
NM_001323892.1 | 3983 | Intron | NP_001310821.1 | |||
NM_001323893.1 | 3983 | Intron | NP_001310822.1 | |||
NM_001323899.1 | 3983 | Intron | NP_001310828.1 | |||
NM_181806.3 | 3983 | Intron | NP_861522.2 | |||
XM_017007740.1 | 3983 | UTR 3 | XP_016863229.1 | |||
XM_017007741.1 | 3983 | Intron | XP_016863230.1 | |||
XM_017007742.1 | 3983 | Intron | XP_016863231.1 | |||
XM_017007743.1 | 3983 | UTR 3 | XP_016863232.1 | |||
XM_017007744.1 | 3983 | Intron | XP_016863233.1 |
KIAA1211 - KIAA1211 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |