Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGATCACTAATCACTTGGAAATTAG[A/G]AATTAGGAAGAAATAGCTACTTAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601603 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C5orf58 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C5orf58 - chromosome 5 open reading frame 58 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001102609.2 | Intron | NP_001096079.1 | ||||
NM_001305393.1 | Intron | NP_001292322.1 | ||||
NM_001305394.1 | Intron | NP_001292323.1 | ||||
NM_001305395.1 | Intron | NP_001292324.1 | ||||
NM_001305396.1 | Intron | NP_001292325.1 | ||||
NM_001305397.1 | Intron | NP_001292326.1 | ||||
XM_011534432.2 | Intron | XP_011532734.1 | ||||
XM_017009029.1 | Intron | XP_016864518.1 | ||||
XM_017009030.1 | Intron | XP_016864519.1 | ||||
XM_017009031.1 | Intron | XP_016864520.1 | ||||
XM_017009032.1 | Intron | XP_016864521.1 | ||||
XM_017009033.1 | Intron | XP_016864522.1 |
LCP2 - lymphocyte cytosolic protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |