Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTCTTAAGGTAACTTACTCTGTTG[C/T]TGAATCATTGTATTCAGTGATGGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607265 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLINT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CLINT1 - clathrin interactor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195555.1 | 1729 | Silent Mutation | CAA,CAG | Q,Q 526 | NP_001182484.1 | |
NM_001195556.1 | 1729 | Silent Mutation | CAA,CAG | Q,Q 508 | NP_001182485.1 | |
NM_014666.3 | 1729 | Silent Mutation | CAA,CAG | Q,Q 508 | NP_055481.1 | |
XM_011534701.2 | 1729 | Silent Mutation | CAA,CAG | Q,Q 526 | XP_011533003.1 | |
XM_017010087.1 | 1729 | Silent Mutation | CAA,CAG | Q,Q 508 | XP_016865576.1 | |
XM_017010088.1 | 1729 | Intron | XP_016865577.1 |