Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCATAATAATTTAAAAAAAAAAT[C/T]CCTGAAAGATCCAAATTGAATAACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 151430 | ||||||||||||||||||||
Literature Links: |
BCL2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BCL2 - BCL2, apoptosis regulator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000633.2 | 5183 | UTR 3 | NP_000624.2 | |||
NM_000657.2 | 5183 | Intron | NP_000648.2 | |||
XM_011526135.2 | 5183 | Intron | XP_011524437.1 | |||
XM_017025917.1 | 5183 | Intron | XP_016881406.1 |