Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAATGGAACCTGGATGCCATGTAAA[C/T]GCAGTAAAAGAAACAGGTTTTGTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605164 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HDAC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HDAC2 - histone deacetylase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001527.3 | 2533 | UTR 3 | NP_001518.3 | |||
XM_011535788.1 | 2533 | Intron | XP_011534090.1 | |||
XM_017010799.1 | 2533 | Intron | XP_016866288.1 |