Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTCTTCAAGGAATCTTCTCGGCTG[G/T]AAAGACTTGAGAAGCTTGTCTCCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612647 | ||||||||||||||||||||
Literature Links: |
RSPH4A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RSPH4A - radial spoke head 4 homolog A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZUFSP - zinc finger with UFM1 specific peptidase domain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145062.2 | 2370 | Missense Mutation | NP_659499.2 | |||
XM_011535565.2 | 2370 | Missense Mutation | XP_011533867.1 | |||
XM_011535567.1 | 2370 | Missense Mutation | XP_011533869.1 | |||
XM_017010393.1 | 2370 | Intron | XP_016865882.1 | |||
XM_017010394.1 | 2370 | Missense Mutation | XP_016865883.1 | |||
XM_017010395.1 | 2370 | Missense Mutation | XP_016865884.1 | |||
XM_017010396.1 | 2370 | Missense Mutation | XP_016865885.1 |