Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGCAGCACCTGACATACTCTTCTT[A/G]TTTCATAGGCAAAGTACTAGCTCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604674 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HEY2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HEY2 - hes related family bHLH transcription factor with YRPW motif 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012259.2 | Intron | NP_036391.1 | ||||
XM_017010627.1 | Intron | XP_016866116.1 | ||||
XM_017010628.1 | Intron | XP_016866117.1 | ||||
XM_017010629.1 | Intron | XP_016866118.1 |
LOC105377986 - uncharacterized LOC105377986 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |