Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACACCCACCTGAATAAATTCTGCAC[A/C]CAGTCTGGCAAACATTTATTGACTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605620 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL20RA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IL20RA - interleukin 20 receptor subunit alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278722.1 | 2204 | UTR 3 | NP_001265651.1 | |||
NM_001278723.1 | 2204 | UTR 3 | NP_001265652.1 | |||
NM_001278724.1 | 2204 | UTR 3 | NP_001265653.1 | |||
NM_014432.3 | 2204 | UTR 3 | NP_055247.3 | |||
XM_006715506.2 | 2204 | UTR 3 | XP_006715569.1 | |||
XM_011535904.2 | 2204 | UTR 3 | XP_011534206.1 | |||
XM_017010954.1 | 2204 | UTR 3 | XP_016866443.1 | |||
XM_017010955.1 | 2204 | UTR 3 | XP_016866444.1 |
NHEG1 - neuroblastoma highly expressed 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |