Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGGGATCGCGGAAAGTGATGGCTG[T/G]CGTCCCGGCGTCTCTCTCAGGACAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616332 MIM: 614917 | ||||||||||||||||||||
Literature Links: |
ARMT1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARMT1 - acidic residue methyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286562.1 | 296 | UTR 5 | NP_001273491.1 | |||
NM_024573.2 | 296 | Missense Mutation | GGC,GTC | G,V 3 | NP_078849.1 |
RMND1 - required for meiotic nuclear division 1 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271937.1 | 296 | Intron | NP_001258866.1 | |||
NM_017909.3 | 296 | Intron | NP_060379.2 | |||
XM_005267040.3 | 296 | Intron | XP_005267097.1 | |||
XM_017010988.1 | 296 | Intron | XP_016866477.1 |