Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGATGAGGTCATTCAACCAGGTTTC[A/G]TCAGCCCCAGGTTAGTCTTTTTTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611044 | ||||||||||||||||||||
Literature Links: |
LIN28B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LIN28B - lin-28 homolog B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004317.3 | 157 | Intron | NP_001004317.1 | |||
XM_006715477.2 | 157 | Silent Mutation | TCA,TCG | S,S 19 | XP_006715540.2 | |
XM_011535818.2 | 157 | Silent Mutation | TCA,TCG | S,S 8 | XP_011534120.1 |
LIN28B-AS1 - LIN28B antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |