Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTATTCTTAGGGTACATGCTGGG[A/T]ACTGAGGGATGAAGTATATGCATAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608126 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
WDHD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
WDHD1 - WD repeat and HMG-box DNA binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008396.2 | 5887 | UTR 3 | NP_001008397.1 | |||
NM_007086.3 | 5887 | UTR 3 | NP_009017.1 | |||
XM_006720012.2 | 5887 | UTR 3 | XP_006720075.1 | |||
XM_011536373.2 | 5887 | Intron | XP_011534675.1 | |||
XM_017020940.1 | 5887 | Intron | XP_016876429.1 | |||
XM_017020941.1 | 5887 | UTR 3 | XP_016876430.1 |