Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCTTTTAGTGTGCGAGCAACCAC[A/G]GGAACTTCTGAAAGTTTTCAAACAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607384 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TIMM9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TIMM9 - translocase of inner mitochondrial membrane 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TOMM20L - translocase of outer mitochondrial membrane 20 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207377.2 | 401 | Missense Mutation | CAG,CGG | Q,R 108 | NP_997260.1 | |
XM_011536742.2 | 401 | Missense Mutation | CAG,CGG | Q,R 116 | XP_011535044.1 | |
XM_011536743.2 | 401 | Missense Mutation | CAG,CGG | Q,R 116 | XP_011535045.1 | |
XM_011536744.2 | 401 | Missense Mutation | CAG,CGG | Q,R 66 | XP_011535046.1 |